missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-buten-2-one, +98%, ACROS Organics™

Brand:  Acros Organics 385592500

Code : 29

Additional Details : CAS Number : 122-57-6 Weight : 0.30000kg


Quantity 250g
Packaging Glass bottle

 View more versions of this product

Product Code. 10510184

  • £12.90 / 250g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

4-Phenyl-3-buten-2-one
98.0
≥98.0%
C10H10O
146.19
122-57-6
100.0
98+%
C6H5CH=CHCOCH3
MFCD00008779

Specifications

4-Phenyl-3-buten-2-one
250g
98.0
Authentic
Low Melting Solid
98+%
C6H5CH=CHCOCH3
MFCD00008779
Pale yellow to yellow
122-57-6
100.0
Glass bottle
≥98.0%
C10H10O
146.19
15, 1140
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Phenyl-3-buten-2-one

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS