missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-thiazoleacetonitrile, 97%, Thermo Scientific™

Catalog Number 15492449
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
41381-89-9
C11H8N2S
200.26
MFCD00551791
SOFCSIBVJROWGI-UHFFFAOYSA-N
2-4-phenylthiazol-2-yl acetonitrile, 2-4-phenyl-1,3-thiazol-2-yl acetonitrile, 4-phenyl-1,3-thiazol-2-yl acetonitrile, 2-cyanomethyl-4-phenylthiazole, 4-phenyl-thiazol-2-yl-acetonitrile, 2-thiazoleacetonitrile, 4-phenyl, 2-4-phenyl-1,3-thiazol-2-yl ethanenitrile, maybridge1_006883, cambridge id 5156617, 4-phenyl-2-cyanomethylthiazole
704539
2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
N#CCC1=NC(=CS1)C1=CC=CC=C1
This item is not returnable. View return policy
41381-89-9
C11H8N2S
200.26
MFCD00551791
SOFCSIBVJROWGI-UHFFFAOYSA-N
2-4-phenylthiazol-2-yl acetonitrile, 2-4-phenyl-1,3-thiazol-2-yl acetonitrile, 4-phenyl-1,3-thiazol-2-yl acetonitrile, 2-cyanomethyl-4-phenylthiazole, 4-phenyl-thiazol-2-yl-acetonitrile, 2-thiazoleacetonitrile, 4-phenyl, 2-4-phenyl-1,3-thiazol-2-yl ethanenitrile, maybridge1_006883, cambridge id 5156617, 4-phenyl-2-cyanomethylthiazole
704539
2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
N#CCC1=NC(=CS1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 250 mg
UN Number UN3439
Formula Weight 200.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Phenyl-2-thiazoleacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'