missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, Tocris Bioscience™

DA release inhibitor

Brand:  Tocris (Bio-Techne) 073010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13457923

  • £106.65 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Inhibitor of release of dopamine induced by methamphetamine.
Specifications

Chemical Identifiers

6109-35-9
245.75
4-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, 4-phenyl-1,2,3,4-tetrahydroisoquinoline hcl, 1,2,3,4-tetrahydro-4-phenyl-isoquinoline hydrochloride
4-phenyl-1,2,3,4-tetrahydroisoquinoline;hydrochloride
C15H16ClN
YYCOJKPDBCMVPP-UHFFFAOYSA-N
12999296
C1C(C2=CC=CC=C2CN1)C3=CC=CC=C3.Cl

Specifications

4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
6109-35-9
4-phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, 4-phenyl-1,2,3,4-tetrahydroisoquinoline hcl, 1,2,3,4-tetrahydro-4-phenyl-isoquinoline hydrochloride
C1C(C2=CC=CC=C2CN1)C3=CC=CC=C3.Cl
245.75
245.75
10mg
C15H16ClN
YYCOJKPDBCMVPP-UHFFFAOYSA-N
4-phenyl-1,2,3,4-tetrahydroisoquinoline;hydrochloride
12999296
>98%
Videos