Learn More
Thermo Scientific Maybridge (4-Phenoxyphenyl)methanol, 1g, 97%
Brand: Thermo Scientific Maybridge CC53709DA
(4-Phenoxyphenyl)methanol, 1g, 97%
Code : 31
Additional Details : CAS Number : 2215-78-3 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC53709DA
(4-Phenoxyphenyl)methanol, 1g, 97%
Code : 31
Additional Details : CAS Number : 2215-78-3 Weight : 0.05100kg