Learn More
Thermo Scientific Maybridge 4-(Phenoxymethyl)benzylamine, 1g, 97%
Brand: Thermo Scientific Maybridge CC63713DA
4-(Phenoxymethyl)benzylamine, 1g, 97%
Code : 31
Additional Details : CAS Number : 872283-78-8 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III