missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-4391446
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1002727-89-0
C13H11ClO3S
282.74
ITLWVYCIZYWFNW-UHFFFAOYSA-N
4-phenoxymethyl benzenesulfonyl chloride, 4-phenoxymethyl benzene-1-sulfonyl chloride
43811043
This item is not returnable. View return policy
1002727-89-0
C13H11ClO3S
282.74
ITLWVYCIZYWFNW-UHFFFAOYSA-N
4-phenoxymethyl benzenesulfonyl chloride, 4-phenoxymethyl benzene-1-sulfonyl chloride
43811043
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 282.75
Percent Purity 97%
Chemical Name or Material 4-(phenoxymethyl)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'