missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzenesulfonyl chloride, 97%, Acros Organics

Brand:  Acros Organics 450350050

Code : NEW

Additional Details : CAS Number : 1623-92-3 Weight : 0.01000kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 15289706

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

4-Phenoxybenzenesulfonyl chloride
97%
C12H9ClO3S
268.72

Specifications

4-Phenoxybenzenesulfonyl chloride
1623-92-3
97%
268.72
5g
97%
C12H9ClO3S
43.0° to 45.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Phenoxybenzenesulfonyl chloride, 97%

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.