missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzaldehyde, 98%, ACROS Organics™

£47.26 - £156.13

Chemical Identifiers

CAS 67-36-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00003383
InChI Key QWLHJVDRPZNVBS-UHFFFAOYSA-N
Synonym p-phenoxybenzaldehyde, 4-formyldiphenyl ether, benzaldehyde, p-phenoxy, benzaldehyde, 4-phenoxy, 4-phenoxy-benzaldehyde, 4-phenyloxy benzaldehyde, diphenyl ether 4-carboxaldehyde, zlchem 548, 4-formyldiphenylether, 4-phenoxy benzaldehyde
PubChem CID 66139
IUPAC Name 4-phenoxybenzaldehyde
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10032490
SDS View Documents
Acros Organics
198930050
5g Glass bottle
£47.26
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10338290
SDS View Documents
Acros Organics
198930250
25g Glass bottle
£156.13
25g
View alternative products
Save Up to 
»
Estimated Shipment
24-05-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67-36-7
198.22
QWLHJVDRPZNVBS-UHFFFAOYSA-N
66139
C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
C13H10O2
MFCD00003383
p-phenoxybenzaldehyde, 4-formyldiphenyl ether, benzaldehyde, p-phenoxy, benzaldehyde, 4-phenoxy, 4-phenoxy-benzaldehyde, 4-phenyloxy benzaldehyde, diphenyl ether 4-carboxaldehyde, zlchem 548, 4-formyldiphenylether, 4-phenoxy benzaldehyde
4-phenoxybenzaldehyde

Specifications

4-Phenoxybenzaldehyde
>112°C
Glass bottle
1.132
Colorless to Yellow
5g
67-36-7
100.0
C13H10O2
MFCD00003383
QWLHJVDRPZNVBS-UHFFFAOYSA-N
4-phenoxybenzaldehyde
66139
Liquid After Melting
1.1320g/mL
Authentic
1.6100 to 1.612
185°C (14.0mmHg)
24°C to 25°C
98%
97.5
97.5% min. (GC)
C6H5OC6H4CHO
p-phenoxybenzaldehyde, 4-formyldiphenyl ether, benzaldehyde, p-phenoxy, benzaldehyde, 4-phenoxy, 4-phenoxy-benzaldehyde, 4-phenyloxy benzaldehyde, diphenyl ether 4-carboxaldehyde, zlchem 548, 4-formyldiphenylether, 4-phenoxy benzaldehyde
C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
198.22
198.22
98%
SDS