missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Phenoxyaniline, 97%

£19.79 - £173.00

Chemical Identifiers

CAS 139-59-3
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00007862
InChI Key WOYZXEVUWXQVNV-UHFFFAOYSA-N
Synonym p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether
PubChem CID 8764
IUPAC Name 4-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418706
SDS View Documents
Alfa Aesar™
A16048.14
25g
£19.79
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
11428706
SDS View Documents
Alfa Aesar™
A16048.22
100g
£46.11
100g
View alternative products
Save Up to 
»
In stock, ready to ship: 5 Please to purchase this item. Need a web account? Register with us today!
11438706
SDS View Documents
Alfa Aesar™
A16048.36
500g
£173.00
500g
View alternative products
Save Up to 
»
In stock, ready to ship: 11 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

139-59-3
185.226
WOYZXEVUWXQVNV-UHFFFAOYSA-N
8764
C1=CC=C(C=C1)OC2=CC=C(C=C2)N
C12H11NO
MFCD00007862
p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether
4-phenoxyaniline

Specifications

4-Phenoxyaniline
82°C to 86°C
139-59-3
MFCD00007862
777708
WOYZXEVUWXQVNV-UHFFFAOYSA-N
4-phenoxyaniline
8764
97%
140°C (2mmHg)
25g
C12H11NO
UN3077
p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether
C1=CC=C(C=C1)OC2=CC=C(C=C2)N
185.226
185.23
SDS