missing translation for 'onlineSavingsMsg'
Learn More

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, Thermo Scientific Chemicals

Catalog Number p-180677
Click to view available options
Quantity:
5 g
100 g
This item is not returnable. View return policy
10580-19-5, 825-90-1
C6H5O4S
173.16
MFCD00044734,MFCD00150724
FEPBITJSIHRMRT-UHFFFAOYSA-M
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
23666329
OC1=CC=C(C=C1)S([O-])(=O)=O
This item is not returnable. View return policy
10580-19-5, 825-90-1
C6H5O4S
173.16
MFCD00044734,MFCD00150724
FEPBITJSIHRMRT-UHFFFAOYSA-M
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
23666329
OC1=CC=C(C=C1)S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color Green to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H4SO3Na·2H2O
Quantity 5 g
Solubility Information Solubility in water: 24g/100mL.
IUPAC Name sodium;4-hydroxybenzenesulfonate;dihydrate
Formula Weight 232.18
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Phenolsulfonic acid, sodium salt dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenolsulfonic acid, sodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'