missing translation for 'onlineSavingsMsg'
Learn More

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, ACROS Organics™

Brand:  Acros Organics 123985000

Code : 29

Additional Details : CAS Number : 825-90-1 Weight : 0.55000kg


Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10428090

  • £109.40 / 500g


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Phenolsulfonic acid, sodium salt dihydrate
825-90-1
100.0
99%
HOC6H4SO3Na·2H2O
MFCD00150724
99%
98.5
99%
C6H5NaO4S·2H2O
232.18

Specifications

4-Phenolsulfonic acid, sodium salt dihydrate
99%
98.5
Authentic
Powder
99%
HOC6H4SO3Na·2H2O
MFCD00150724
13 to 17% (K.F.)
> 300.0°C
500g
825-90-1
100.0
Plastic bottle
99%
C6H5NaO4S·2H2O
232.18
Solubility in water: 24g/100mL.
Pale yellow to beige to gold to green
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Phenolsulfonic acid, sodium salt dihydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS