missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-1-ol, 95%, Thermo Scientific Chemicals

Catalog Number p-7080745
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5390-04-5
MFCD00002974
This item is not returnable. View return policy
5390-04-5
MFCD00002974

4-Pentyn-1-ol is widely utilized in ring-opening polymerization reactions. It plays an important role as a starting reagent in stereo selective total synthesis of antimicrobial marine metabolites, ieodomycin A and B. It is also used in the preparation of 3-pent-4-ynyloxy phthalonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentyn-1-ol is widely utilized in ring-opening polymerization reactions. It plays an important role as a starting reagent in stereo selective total synthesis of antimicrobial marine metabolites, ieodomycin A and B. It is also used in the preparation of 3-pent-4-ynyloxy phthalonitrile.

Solubility
Miscible with water.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Density 0.907
Boiling Point 153°C to 155°C
Flash Point 62°C (143°F)
Molecular Formula C9H9BrO3
Linear Formula HC≡C(CH2)3OH
Refractive Index 1.445
Quantity 5 g
Beilstein 1736712
Synonym 6-bromoveratraldehyde, 6-bromo-3,4-dimethoxybenzaldehyde, benzaldehyde, 2-bromo-4,5-dimethoxy, 6-bromo veratraldehyde, 2-bromo-4,5-dimethoxybenzalehyde, 4,5-dimethoxy-2-bromobenzaldehyde, 6-bromoveratral, pubchem8209, veratraldehyde, 6-bromo, acmc-2097hh
Solubility Information Miscible with water.
InChI Key UQQROBHFUDBOOK-UHFFFAOYSA-N
SMILES COC1=C(C=C(C(=C1)C=O)Br)OC
IUPAC Name 2-bromo-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol) 245.07
PubChem CID 79351
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 4-Pentyn-1-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'