missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzonitrile, 97%, Thermo Scientific™

Catalog Number p-2702102
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10270-29-8
C12H15N
173.26
MFCD00218832
LGPQFJXPPKNHJU-UHFFFAOYSA-N
4-n-pentylbenzonitrile, 4-pentylbenzenecarbonitrile, p-amylbenzonitrile, p-n-pentylbenzonitrile, benzonitrile, 4-pentyl
264951
4-pentylbenzonitrile
CCCCCC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
10270-29-8
C12H15N
173.26
MFCD00218832
LGPQFJXPPKNHJU-UHFFFAOYSA-N
4-n-pentylbenzonitrile, 4-pentylbenzenecarbonitrile, p-amylbenzonitrile, p-n-pentylbenzonitrile, benzonitrile, 4-pentyl
264951
4-pentylbenzonitrile
CCCCCC1=CC=C(C=C1)C#N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)4C6H4CN
Quantity 1 g
Formula Weight 173.26
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'