missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-2702098
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
73948-18-2
C11H15ClO2S
246.75
MFCD00173759
XDWXZRVWJHEWMT-UHFFFAOYSA-N
4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
144625
CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
73948-18-2
C11H15ClO2S
246.75
MFCD00173759
XDWXZRVWJHEWMT-UHFFFAOYSA-N
4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
144625
CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CH3(CH2)4C6H4SO2Cl
Quantity 1 g
IUPAC Name 4-pentylbenzenesulfonyl chloride
Formula Weight 246.76
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'