Learn More
4-Penten-1-ol, 98+%, Thermo Scientific Chemicals
Description
4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.
Solubility
Solubility in water: 57 g/L (25°C)
Notes
Protect against electrostatic charges. Store away from oxidizing agents. Use only in a chemical fume hood. Store at 2-8°C.
Specifications
Density | 0.84 |
Boiling Point | 134°C to 137°C |
Flash Point | 43°C (109°F) |
Refractive Index | 1.43 |
Quantity | 5 g |
UN Number | UN1987 |
Beilstein | 1560163 |
Formula Weight | 86.13 |
Percent Purity | ≥98% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only