missing translation for 'onlineSavingsMsg'
Learn More

4-P-PDOT, Tocris Bioscience™

MT2 antagonist

Brand:  Tocris (Bio-Techne) 1034/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438353

  • £149.94 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Melatonin receptor antagonist, >300-fold selective for the MT2 (vs MT1) subtype.
Specifications

Chemical Identifiers

134865-74-0
279.383
4p-pdot, 4-p-pdot, n-4alpha-phenyl-1,2,3,4-tetrahydronaphthalene-2alpha-yl propanamide
N-[(2R,4R)-4-phenyl-1,2,3,4-tetrahydronaphthalen-2-yl]propanamide
C19H21NO
RCYLUNPFECYGDW-FUHWJXTLSA-N
56935196
CCC(=O)NC1CC(C2=CC=CC=C2C1)C3=CC=CC=C3

Specifications

4-P-PDOT
134865-74-0
4p-pdot, 4-p-pdot, n-4alpha-phenyl-1,2,3,4-tetrahydronaphthalene-2alpha-yl propanamide
CCC(=O)NC1CC(C2=CC=CC=C2C1)C3=CC=CC=C3
279.383
279.38
10mg
C19H21NO
RCYLUNPFECYGDW-FUHWJXTLSA-N
N-[(2R,4R)-4-phenyl-1,2,3,4-tetrahydronaphthalen-2-yl]propanamide
56935196
>99%
Videos
Documents