missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 95%, free radical, ACROS Organics™

Brand:  Acros Organics 176860050

Code : 29

Additional Details : CAS Number : 2896-70-0 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10367450

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Polymerization reagent
Specifications

Chemical Identifiers

4-Oxo-2,2,6,6-tetramethylpiperidinooxy
2896-70-0
100.0
95%
170.23
2,2,6,6-Tetramethyl-4-oxopiperidinooxy, 4-Oxo-TEMPO
95%
94.0
≥94%
C9H16NO2
MFCD00006187

Specifications

4-Oxo-2,2,6,6-tetramethylpiperidinooxy
2896-70-0
100.0
Needles, Crystals or Crystalline Powder
95%
170.23
Glass bottle
Orange
95%
94.0
Authentic
≥94%
C9H16NO2
MFCD00006187
2,2,6,6-Tetramethyl-4-oxopiperidinooxy, 4-Oxo-TEMPO
5g
Videos
SDS