missing translation for 'onlineSavingsMsg'
Learn More

4-Octyne, 98+%, Thermo Scientific Chemicals

Catalog Number p-3758617
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1942-45-6
C8H14
110.2
MFCD00009471
GZTNBKQTTZSQNS-UHFFFAOYSA-N
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
16029
oct-4-yne
CCCC#CCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1942-45-6
C8H14
110.2
MFCD00009471
GZTNBKQTTZSQNS-UHFFFAOYSA-N
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
16029
oct-4-yne
CCCC#CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -103.0°C
Color Colorless to Yellow
Density 0.7500g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C≡CCH2CH2CH3
Refractive Index 1.4240 to 1.4260
Quantity 10 mL
Beilstein 01, III, 1006
Specific Gravity 0.75
Formula Weight 110.2
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Octyne
Show More Show Less
compliance-icons
Product Identifier
  • 4-Octyne
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'