missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiophenol, 80%, Tech., ACROS Organics™

Brand:  Acros Organics 336200250

Code : 29

Additional Details : CAS Number : 1849-36-1 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10693332

  • £264.80 / 25g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

4-Nitrothiophenol
1849-36-1
100.0
80%
C6H5NO2S
155.2
4-Nitrobenzenethiol
95%
75.0
Technical Grade
80%
SHC6H4NO2
MFCD00007343

Specifications

4-Nitrothiophenol
1849-36-1
100.0
Glass bottle
Technical Grade
Crystalline Powder and/or Chunks
80%
C6H5NO2S
155.2
06, 339
95%
75.0
Authentic
72.0° to 80.0°C
Yellow
80%
25g
SHC6H4NO2
MFCD00007343
4-Nitrobenzenethiol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Nitrothiophenol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS