missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiophenol, 80%, Tech., ACROS Organics™

Brand:  Acros Organics 336200050

Code : 29

Additional Details : CAS Number : 1849-36-1 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10400442

  • £83.00 / 5g


Call for latest availability
+44 (0)1509 555 500
Add to basket


Specifications

Chemical Identifiers

4-Nitrothiophenol
1849-36-1
100.0
80%
C6H5NO2S
155.2
4-Nitrobenzenethiol
95%
75.0
Technical Grade
80%
SHC6H4NO2
MFCD00007343

Specifications

4-Nitrothiophenol
1849-36-1
100.0
Glass bottle
Technical Grade
Crystalline Powder and/or Chunks
80%
C6H5NO2S
155.2
06, 339
95%
75.0
Authentic
72.0° to 80.0°C
Yellow
80%
5g
SHC6H4NO2
MFCD00007343
4-Nitrobenzenethiol
Videos
SDS