missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-4509629
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
610-27-5
C8H5NO6
211.13
SLBQXWXKPNIVSQ-UHFFFAOYSA-N
4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8
69121
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
610-27-5
C8H5NO6
211.13
SLBQXWXKPNIVSQ-UHFFFAOYSA-N
4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8
69121
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 09,828
Formula Weight 211.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'