missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic acid, 99%, ACROS Organics™

Brand:  Acros Organics 416125000

Code : 29

Additional Details : CAS Number : 610-27-5 Weight : 0.55000kg

Packaging Glass bottle
Quantity 500g

 View more versions of this product

Product Code. 11975051

  • £1478.20 / 500g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

4-Nitrophthalic acid
99%
211.13
99%
C8H5NO6

Specifications

4-Nitrophthalic acid
Authentic
Crystalline Powder
99%
Yellow to beige
165.0°C
610-27-5
Glass bottle
99%
C8H5NO6
211.13
500g
Videos
SDS