missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenylglyoxal hydrate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
As important industrial compounds and are mainly used as solvents. A new method for arginase assay
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
As important industrial compounds and are mainly used as solvents. A new method for arginase assay
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 96°C to 98°C |
Color | Cream |
Quantity | 10 g |
Formula Weight | 179.13 (anhy.) |
Percent Purity | 97% |
Physical Form | Crystalline Powder |
Chemical Name or Material | 4-Nitrophenylglyoxal hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'