missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7045483
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1492-30-4
C22H35NO4
377.525
MFCD00047732
LVZSQWIWCANHPF-UHFFFAOYSA-N
4-nitrophenyl palmitate, p-nitrophenyl palmitate, hexadecanoic acid 4-nitrophenyl ester, 4-nitrophenyl hexadecanoate, hexadecanoic acid, 4-nitrophenyl ester, 4-nitrophenylpalmitate, paranitrophenyl palmitate, para-nitrophenyl palmitate, p-nitrophenyl hexadecanoate, 4-nitrophenyl palmitate #
73891
CHEBI:85645
(4-nitrophenyl) hexadecanoate
CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
1492-30-4
C22H35NO4
377.525
MFCD00047732
LVZSQWIWCANHPF-UHFFFAOYSA-N
4-nitrophenyl palmitate, p-nitrophenyl palmitate, hexadecanoic acid 4-nitrophenyl ester, 4-nitrophenyl hexadecanoate, hexadecanoic acid, 4-nitrophenyl ester, 4-nitrophenylpalmitate, paranitrophenyl palmitate, para-nitrophenyl palmitate, p-nitrophenyl hexadecanoate, 4-nitrophenyl palmitate #
73891
CHEBI:85645
(4-nitrophenyl) hexadecanoate
CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]

The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

Solubility
Insoluble in water. Soluble in CHCl3.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C
TRUSTED_SUSTAINABILITY
Melting Point 65°C to 66°C
Quantity 1 g
Beilstein 1891754
Solubility Information Insoluble in water. Soluble in CHCl3.
Formula Weight 377.53
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl palmitate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'