missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-2702058
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
100-28-7
C7H4N2O3
164.12
GFNKTLQTQSALEJ-UHFFFAOYSA-N
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
66012
1-isocyanato-4-nitrobenzene
C1=CC(=CC=C1N=C=O)[N+](=O)[O-]
This item is not returnable. View return policy
100-28-7
C7H4N2O3
164.12
GFNKTLQTQSALEJ-UHFFFAOYSA-N
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
66012
1-isocyanato-4-nitrobenzene
C1=CC(=CC=C1N=C=O)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 62.0°C
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NCO
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in benzene and toluene
Formula Weight 164.12
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Nitrophenyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'