missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Disodium Orthophosphate, Pure, Fisher Chemical™

Catalog Number p-7073344
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
333338-18-4
C6H16NNa2O12P
371.14
MFCD00007319
KAKKHKRHCKCAGH-UHFFFAOYSA-L
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
This item is not returnable. View return policy
333338-18-4
C6H16NNa2O12P
371.14
MFCD00007319
KAKKHKRHCKCAGH-UHFFFAOYSA-L
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

4-Nitrophenyl Disodium Orthophosphate, Pure, Fisher Chemical, Quantity: 25g, CAS: 333338-18-4, White, Melting Point: 300 deg.C, Molecular Formula: C6H4NNa2O6P, Molecular Weight: 263.052, Packaging: Plastic powder jar

TRUSTED_SUSTAINABILITY
Melting Point 300°C
Color White
Physical Form Powder Solid
Quantity 5 g
Formula Weight 371.13g/mol
Grade Pure
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'