missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl-beta-D-glucuronide, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.05% max. (p-Nitrophenol) (HPLC) |
Packaging | Glass bottle |
Quantity | 50 mg |
Specific Rotation | −107° to −113° (20°C, 589nm) (c=1, H2O) |
Solubility Information | (1% in water) Clear colorless to light yellow |
Formula Weight | 315.24 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'