missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Catalog Number p-174533
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3459-18-5
C14H18N2O8
342.30
MFCD00063696
OMRLTNCLYHKQCK-JQXIRTQVNA-N
4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide
102416
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
3459-18-5
C14H18N2O8
342.30
MFCD00063696
OMRLTNCLYHKQCK-JQXIRTQVNA-N
4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide
102416
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 210.0°C to 212.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 18, V,11, 102
Specific Rotation Condition −16° (20°C c=0.5,water)
Specific Rotation − 16.00
Solubility Information (0.5% in water) Clear colorless to light yellow
Formula Weight 342.29
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material 4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'