missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenethyl bromide, 98%, Thermo Scientific Chemicals

Catalog Number 10541401
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5339-26-4
C8H8BrNO2
230.06
MFCD00007386
NTURQZFFJDCTMZ-UHFFFAOYSA-N
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
79266
1-(2-bromoethyl)-4-nitrobenzene
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
This item is not returnable. View return policy
5339-26-4
C8H8BrNO2
230.06
MFCD00007386
NTURQZFFJDCTMZ-UHFFFAOYSA-N
4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
79266
1-(2-bromoethyl)-4-nitrobenzene
C1=CC(=CC=C1CCBr)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 67°C to 70°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4CH2CH2Br
Quantity 25 g
Formula Weight 230.06
Percent Purity 98%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 4-Nitrophenethyl bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrophenethyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'