missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitronicotinic acid N-oxide, 97%

Brand:  Alfa Aesar™ L20049.03

UNSPSC : 12352005

Code : NEW

Additional Details : CAS Number : 1078-05-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11364466

  • £20.67 / 1g


Estimated Shipment date
from Supplier 11-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

4-Nitronicotinic acid N-oxide
C6H4N2O5
MFCD03426925
4-Nitropyridine-3-carboxylic acid N-oxide
1078-05-3
184.11
176825

Specifications

4-Nitronicotinic acid N-oxide
≈170°C
1078-05-3
C6H4N2O5
MFCD03426925
4-Nitropyridine-3-carboxylic acid N-oxide
Odorless
1g
97%
184.11
176825
Documentation

Technical Documents