missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitronicotinic acid N-oxide, 97%

Brand:  Alfa Aesar™ L20049.06

Code : NEW

Additional Details : CAS Number : 1078-05-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11374466

  • £68.90 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Nitronicotinic acid N-oxide
C6H4N2O5
MFCD03426925
4-Nitropyridine-3-carboxylic acid N-oxide
1078-05-3
184.11
176825

Specifications

4-Nitronicotinic acid N-oxide
≈170°C
1078-05-3
C6H4N2O5
MFCD03426925
4-Nitropyridine-3-carboxylic acid N-oxide
Odorless
5g
97%
184.11
176825
Videos
SDS