missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitroindole, 98%, Thermo Scientific™
Click to view available options
2.5g; Glass bottle
Specifications
CAS | 4769-97-5 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 203°C to 207°C |
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C8H6N2O2 |
MDL Number | MFCD00010056 |
Show More |
Safety and Handling
Product Identifier
- 4-Nitroindole
- Warning
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'