missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitroindole-3-carboxaldehyde, 97%

Brand:  Alfa Aesar™ H63595.MD

Code : NEW

Additional Details : CAS Number : 10553-11-4 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15470369

  • £13.60 / 250mg


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Nitroindole-3-carboxaldehyde
190.16
3-Formyl-4-nitroindole
10553-11-4
MFCD04973979

Specifications

4-Nitroindole-3-carboxaldehyde
189° to 193°C
10553-11-4
97%
MFCD04973979
Yellow
250mg
Solid
190.16
3-Formyl-4-nitroindole
Videos
SDS