missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitrohippuric acid, 98%

Brand:  Alfa Aesar™ A12347.14

Code : NEW

Additional Details : CAS Number : 2645-07-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11459473

  • £133.80 / 25g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Nitrohippuric acid
C9H8N2O5
MFCD00007349
2645-07-0
224.17
N-(4-Nitrobenzoyl)glycine

Specifications

4-Nitrohippuric acid
25g
98%
224.17
N-(4-Nitrobenzoyl)glycine
128° to 131°C
2645-07-0
C9H8N2O5
MFCD00007349
Videos
SDS