missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocinnamaldehyde, predominantly trans, 98%, Thermo Scientific Chemicals

Catalog Number p-7081471
41.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1734-79-8
C9H7NO3
177.159
MFCD00007379
ALGQVMMYDWQDEC-OWOJBTEDSA-N
4-nitrocinnamaldehyde, 3-4-nitrophenyl acrylaldehyde, p-nitrocinnamaldehyde, 2-propenal, 3-4-nitrophenyl, cinnamaldehyde, p-nitro, trans-4-nitrocinnamaldehyde, e-3-4-nitrophenyl acrylaldehyde, 3-4-nitrophenyl-2-propenal, unii-48r8lam7yx, 2e-3-4-nitrophenyl prop-2-enal
5354135
(E)-3-(4-nitrophenyl)prop-2-enal
C1=CC(=CC=C1C=CC=O)[N+](=O)[O-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1734-79-8
C9H7NO3
177.159
MFCD00007379
ALGQVMMYDWQDEC-OWOJBTEDSA-N
4-nitrocinnamaldehyde, 3-4-nitrophenyl acrylaldehyde, p-nitrocinnamaldehyde, 2-propenal, 3-4-nitrophenyl, cinnamaldehyde, p-nitro, trans-4-nitrocinnamaldehyde, e-3-4-nitrophenyl acrylaldehyde, 3-4-nitrophenyl-2-propenal, unii-48r8lam7yx, 2e-3-4-nitrophenyl prop-2-enal
5354135
(E)-3-(4-nitrophenyl)prop-2-enal
C1=CC(=CC=C1C=CC=O)[N+](=O)[O-]

 

Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4'-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2?-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4′-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2′-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Odor Characteristic
Quantity 5 g
Beilstein 1565424
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 4-Nitrocinnamaldehyde, predominantly trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'