missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloroformate, 95%, Thermo Scientific Chemicals

Catalog Number p-3598210
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4457-32-3
C8H6ClNO4
215.59
MFCD00007375
MHSGOABISYIYKP-UHFFFAOYSA-N
4-nitrobenzyl chloroformate, 4-nitrobenzyl carbonochloridate, carbonochloridic acid, 4-nitrophenyl methyl ester, 4-nitrobenzyl chlorocarbonate, p-nitrobenzyl chloroformate, 4-nitrocarbobenzoxychloride, 4-nitrophenyl methyl chloroformate, p-nitrobenzyloxycarbonyl chloride, 4-nitrobenzyloxycarbonyl chloride, chloroformic acid p-nitrobenzyl ester
78205
(4-nitrophenyl)methyl carbonochloridate
C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
4457-32-3
C8H6ClNO4
215.59
MFCD00007375
MHSGOABISYIYKP-UHFFFAOYSA-N
4-nitrobenzyl chloroformate, 4-nitrobenzyl carbonochloridate, carbonochloridic acid, 4-nitrophenyl methyl ester, 4-nitrobenzyl chlorocarbonate, p-nitrobenzyl chloroformate, 4-nitrocarbobenzoxychloride, 4-nitrophenyl methyl chloroformate, p-nitrobenzyloxycarbonyl chloride, 4-nitrobenzyloxycarbonyl chloride, chloroformic acid p-nitrobenzyl ester
78205
(4-nitrophenyl)methyl carbonochloridate
C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 32°C
Flash Point 113°C
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass Bottle
Linear Formula ClCO2CH2C6H4NO2
Refractive Index 1.5520 to 1.556
Quantity 5 g
Formula Weight 215.59
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzyl chloroformate
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrobenzyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'