missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-173007
33.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


31.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
619-72-7
C7H4N2O2
148.12
MFCD00007279
NKJIFDNZPGLLSH-UHFFFAOYSA-N
benzonitrile, 4-nitro, 4-cyanonitrobenzene, p-nitrobenzonitrile, p-cyanonitrobenzene, benzonitrile, p-nitro, 4-nitrobenzenecarbonitrile, 1-nitro-4-cyanobenzene, 4-nitro-benzonitrile, ccris 2328, 4-nitrobenzonitril
12090
4-nitrobenzonitrile
[O-][N+](=O)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
619-72-7
C7H4N2O2
148.12
MFCD00007279
NKJIFDNZPGLLSH-UHFFFAOYSA-N
benzonitrile, 4-nitro, 4-cyanonitrobenzene, p-nitrobenzonitrile, p-cyanonitrobenzene, benzonitrile, p-nitro, 4-nitrobenzenecarbonitrile, 1-nitro-4-cyanobenzene, 4-nitro-benzonitrile, ccris 2328, 4-nitrobenzonitril
12090
4-nitrobenzonitrile
[O-][N+](=O)C1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 144.0°C to 149.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 25 g
Beilstein 09,397
Solubility Information Solubility in water: sparingly soluble
Formula Weight 148.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Nitrobenzonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'