missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-172291
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
98-74-8
C6H4ClNO4S
221.61
MFCD00007445
JXRGUPLJCCDGKG-UHFFFAOYSA-N
4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
7404
4-nitrobenzenesulfonyl chloride
[O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
98-74-8
C6H4ClNO4S
221.61
MFCD00007445
JXRGUPLJCCDGKG-UHFFFAOYSA-N
4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
7404
4-nitrobenzenesulfonyl chloride
[O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 75°C to 80°C
Color Yellow to Brown
Boiling Point 143°C to 144°C (2.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula SO2ClC6H4NO2
Quantity 5 g
Beilstein 11, 72
Solubility Information Solubility in water: insoluble
Formula Weight 221.62
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'