missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals

Catalog Number 10668904
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104-04-1
C8H8N2O3
180.16
MFCD00007303
NQRLPDFELNCFHW-UHFFFAOYSA-N
4'-nitroacetanilide, n-4-nitrophenyl acetamide, 4-nitroacetanilide, p-nitroacetanilide, acetamide, n-4-nitrophenyl, n-acetyl-4-nitroaniline, p-acetamidonitrobenzene, acetanilide, p-nitro, p-nitrophenylacetanilide, n-acetyl-p-nitroaniline
7691
N-(4-nitrophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
104-04-1
C8H8N2O3
180.16
MFCD00007303
NQRLPDFELNCFHW-UHFFFAOYSA-N
4'-nitroacetanilide, n-4-nitrophenyl acetamide, 4-nitroacetanilide, p-nitroacetanilide, acetamide, n-4-nitrophenyl, n-acetyl-4-nitroaniline, p-acetamidonitrobenzene, acetanilide, p-nitro, p-nitrophenylacetanilide, n-acetyl-p-nitroaniline
7691
N-(4-nitrophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 213°C to 216°C
Color Brown-Green or Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C(O)NHC6H4NO2
Quantity 500 g
Merck Index 15, 6667
Solubility Information Solubility in water: 2.2g/L (25°C)
Formula Weight 180.16
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4′-Nitroacetanilide
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Nitroacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'