missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitro-DL-phenylalanine, 98%

Brand:  Alfa Aesar™ H63067.03

Code : NEW

Additional Details : CAS Number : 2922-40-9 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15439548

  • £23.43 / 1g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Nitro-DL-phenylalanine
210.19
UN2811
2922-40-9
MFCD00007384
H-DL-Phe(4-NO2)-OH

Specifications

4-Nitro-DL-phenylalanine
Solid
210.19
UN2811
236° to 237°C (dec.)
2922-40-9
98%
MFCD00007384
H-DL-Phe(4-NO2)-OH
1g
Videos
SDS