missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)phenol, 99%, Thermo Scientific™

Catalog Number 10480921
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
88-30-2
C7H4F3NO3
207.11
MFCD00014714
ZEFMBAFMCSYJOO-UHFFFAOYSA-N
4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
6931
4-nitro-3-(trifluoromethyl)phenol
C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]
This item is not returnable. View return policy
88-30-2
C7H4F3NO3
207.11
MFCD00014714
ZEFMBAFMCSYJOO-UHFFFAOYSA-N
4-nitro-3-trifluoromethyl phenol, lamprecide, lamprecid, 5-hydroxy-2-nitrobenzotrifluoride, dowlap f, 3-trifluoromethyl-4-nitrophenol, phenol, 4-nitro-3-trifluoromethyl, usaf ma-6, caswell no. 890, tfm piscicide
6931
4-nitro-3-(trifluoromethyl)phenol
C1=CC(=C(C=C1O)C(F)(F)F)[N+](=O)[O-]
CAS Min % 98.5
CAS Max % 100.0
Melting Point 72°C to 79°C
Color Yellow
Boiling Point 135°C to 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H3(CF3)OH
Quantity 1 g
Merck Index 15, 9401
Formula Weight 207.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)phenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'