missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitro-3-(trifluoromethyl)anisole, 96%

Brand:  Alfa Aesar™ H61402.14

Code : NEW

Additional Details : CAS Number : 344-39-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15437938

  • £80.67 / 25g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Nitro-3-(trifluoromethyl)anisole
221.13
5-Methoxy-2-nitrobenzotrifluoride; 4-Methoxy-1-nitro-2-(trifluoromethyl)benzene
344-39-8
MFCD00053611

Specifications

4-Nitro-3-(trifluoromethyl)anisole
25g
Crystalline Solid
221.13
5-Methoxy-2-nitrobenzotrifluoride; 4-Methoxy-1-nitro-2-(trifluoromethyl)benzene
Yellow
344-39-8
96%
MFCD00053611
Videos
SDS