missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenylboronic acid, 95%, Thermo Scientific™

Catalog Number 10871671
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
134150-01-9
C9H13BO2
164.01
MFCD00859261
WLCGYIWOKVWFLB-UHFFFAOYSA-N
4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid
4100861
(4-propylphenyl)boronic acid
CCCC1=CC=C(C=C1)B(O)O
This item is not returnable. View return policy
134150-01-9
C9H13BO2
164.01
MFCD00859261
WLCGYIWOKVWFLB-UHFFFAOYSA-N
4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid
4100861
(4-propylphenyl)boronic acid
CCCC1=CC=C(C=C1)B(O)O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3(CH2)2C6H4B(OH)2
Quantity 5 g
Formula Weight 164.01
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Propylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-n-Propylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'