missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Propylphenylboronic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 111.0°C to 114.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)2C6H4B(OH)2 |
Quantity | 1 g |
Formula Weight | 164.01 |
Show More |
Safety and Handling
Product Identifier
- 4-n-Propylphenylboronic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'