missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-n-Propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 98%

Brand:  Alfa Aesar™ H56495.06

Code : NEW

Additional Details : CAS Number : 67589-54-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15402736

  • £198.00 / 5g

Estimated Shipment: 24-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

67589-54-2
332.484
ZAUPPIHFDVKVJM-UHFFFAOYSA-N
105461
CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCCC
C21H32O3
MFCD09751071
cyclohexanecarboxylic acid, 4-pentyl-, 4-propoxyphenyl ester, trans, 4-propoxyphenyl 4-pentylcyclohexane-1-carboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-propoxyphenyl ester, 4-propoxyphenyl 4-pentylcyclohexanecarboxylate, p-propoxyphenyl trans-4-pentylcyclohexanecarboxylate, 4-n-propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 4-pentyl-1-cyclohexanecarboxylic acid 4-propoxyphenyl ester, trans-4-pentylcyclohexanecarboxylic acid 4-propoxyphenyl ester, 4-propoxyphenyl 1s,4r-4-pentylcyclohexane-1-carboxylate, 4alpha-pentyl-1beta-cyclohexanecarboxylic acid 4-propyloxy phenyl ester
(4-propoxyphenyl) 4-pentylcyclohexane-1-carboxylate

Specifications

4-n-Propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate
67589-54-2
MFCD09751071
ZAUPPIHFDVKVJM-UHFFFAOYSA-N
(4-propoxyphenyl) 4-pentylcyclohexane-1-carboxylate
105461
98%
5g
C21H32O3
cyclohexanecarboxylic acid, 4-pentyl-, 4-propoxyphenyl ester, trans, 4-propoxyphenyl 4-pentylcyclohexane-1-carboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-propoxyphenyl ester, 4-propoxyphenyl 4-pentylcyclohexanecarboxylate, p-propoxyphenyl trans-4-pentylcyclohexanecarboxylate, 4-n-propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 4-pentyl-1-cyclohexanecarboxylic acid 4-propoxyphenyl ester, trans-4-pentylcyclohexanecarboxylic acid 4-propoxyphenyl ester, 4-propoxyphenyl 1s,4r-4-pentylcyclohexane-1-carboxylate, 4alpha-pentyl-1beta-cyclohexanecarboxylic acid 4-propyloxy phenyl ester
CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCCC
332.484
332.48
Videos
SDS Documents
Certificates