missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7081861
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
26311-45-5
C12H16O2
192.258
MFCD00002572
CWYNKKGQJYAHQG-UHFFFAOYSA-N
benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
33479
4-pentylbenzoic acid
CCCCCC1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
26311-45-5
C12H16O2
192.258
MFCD00002572
CWYNKKGQJYAHQG-UHFFFAOYSA-N
benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
33479
4-pentylbenzoic acid
CCCCCC1=CC=C(C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Beilstein 2044798
Formula Weight 192.26
Percent Purity 99%
Chemical Name or Material 4-n-Pentylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'