missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7047208
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
121219-12-3
C11H17BO2
192.07
MFCD00995151
UZRMPSOGFATLJE-UHFFFAOYSA-N
4-n-pentylphenylboronic acid, 4-pentylphenyl boronic acid, 4-pentylbenzeneboronic acid, 4-n-pentylbenzeneboronic acid, 4-amylphenylboronic acid, 4-pentylbenzene boronic acid, 4-pentylphenylboronicacid, 4-amylbenzeneboronic acid, boronic acid, 4-pentylphenyl, pubchem16788
4589191
(4-pentylphenyl)boronic acid
CCCCCC1=CC=C(C=C1)B(O)O
This item is not returnable. View return policy
121219-12-3
C11H17BO2
192.07
MFCD00995151
UZRMPSOGFATLJE-UHFFFAOYSA-N
4-n-pentylphenylboronic acid, 4-pentylphenyl boronic acid, 4-pentylbenzeneboronic acid, 4-n-pentylbenzeneboronic acid, 4-amylphenylboronic acid, 4-pentylbenzene boronic acid, 4-pentylphenylboronicacid, 4-amylbenzeneboronic acid, boronic acid, 4-pentylphenyl, pubchem16788
4589191
(4-pentylphenyl)boronic acid
CCCCCC1=CC=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 82°C to 84°C
Quantity 250 mg
Beilstein 4782754
Formula Weight 192.07
Percent Purity 97%
Chemical Name or Material 4-n-Pentylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'