missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4'-n-Pentylacetophenone, 98%

Brand:  Alfa Aesar™ A10725.14

Code : NEW

Additional Details : CAS Number : 37593-02-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11439592

  • £269.70 / 25g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4'-n-Pentylacetophenone
C13H18O
MFCD00043687
37593-02-5
190.29
4'-n-Amylacetophenone

Specifications

4'-n-Pentylacetophenone
25g
98%
190.29
4'-n-Amylacetophenone
132° to 133°C (4mmHg)
37593-02-5
C13H18O
MFCD00043687
Videos
SDS