missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-n-Octyloxyaniline, 98%

Brand:  Alfa Aesar™ H55706.06

Code : NEW

Additional Details : CAS Number : 39905-45-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15492028

  • £45.58 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

4-n-Octyloxyaniline
39905-45-8
221.34
MFCD00025333

Specifications

4-n-Octyloxyaniline
White to brown
5g
Solid
221.34
336°C
35° to 39°C
39905-45-8
98%
MFCD00025333
Videos
SDS