missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Dimethylamino)phenylboronic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-115703
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
28611-39-4
C8H12BNO2
165.00
MFCD01074642
RIIPFHVHLXPMHQ-UHFFFAOYSA-N
4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid, 4-dimethylaminophenyl boronic acid, 4-dimethylamino phenyl boranediol, 4-n,n-dimethylphenylboronic acid
2734344
[4-(dimethylamino)phenyl]boronic acid
CN(C)C1=CC=C(C=C1)B(O)O
This item is not returnable. View return policy
28611-39-4
C8H12BNO2
165.00
MFCD01074642
RIIPFHVHLXPMHQ-UHFFFAOYSA-N
4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid, 4-dimethylaminophenyl boronic acid, 4-dimethylamino phenyl boranediol, 4-n,n-dimethylphenylboronic acid
2734344
[4-(dimethylamino)phenyl]boronic acid
CN(C)C1=CC=C(C=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 227°C
Color Blue-Gray or Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 165
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-(N, N-Dimethylamino)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(N,N-Dimethylamino)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'